Invitrogen
Manufacturer Code:MA535779
Catalog # PIMA535779
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1477 |
Immunogen | A synthesized peptide derived from human Annexin IV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 35-beta calcimedin; 35-beta calcimedin AI265406 AIV Annexin A4 Annexin IV Annexin-4 Anx4 ANXA 4 Anxa4 ANXIV AW106930 BOS_11746 Carbohydrate-binding protein p33/p41 Chromobindin-4 Endonexin I HEL-S-274 Lipocortin IV P32.5 PAP-II PIG28 Placental anticoagulant protein II PP4-X Protein II Xanx-4 ZAP36; 36 kDa zymogen granule membrane-associated protein; Annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); Annexin-4; Carbohydrate-binding protein p33/p41; Chromobindin-4; Endonexin I; epididymis secretory protein Li 274; Lipocortin IV; P32.5; PAP-II; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II; ZAP 36/annexin IV; ZAP36
Gene Aliases: AI265406; AIV; ANX4; ANXA4; AW106930; HEL-S-274; P32.5; PAP-II; PIG28; PP4-X; Xanx-4; ZAP36
UniProt ID: (Human) P09525, (Mouse) P97429
Entrez Gene ID: (Human) 307, (Rat) 79124, (Mouse) 11746
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.