R&D Systems
Manufacturer Code:MAB4146
Catalog # MAB4146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 694624 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Annexin A4 Met1-Asp319 Accession # P09525 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 35-beta calcimedin; AIV annexin A4 Annexin IV annexin IV (placental anticoagulant protein II) annexin-4 ANX4 ANX4Carbohydrate-binding protein p33/p41 ANXA4 Chromobindin-4 chromobindin-435-beta calcimedin DKFZp686H02120 Endonexin I Lipocortin IV MGC75105 P32.5 PAP-II PIG28 Placental anticoagulant protein II PP4-X proliferation-inducing gene 28 proliferation-inducing protein 28 Protein II ZAP36; Annexin A4; Annexin IV; annexin IV (placental anticoagulant protein II); Annexin-4; Carbohydrate-binding protein p33/p41; Chromobindin-4; Endonexin I; epididymis secretory protein Li 274; Lipocortin IV; P32.5; PAP-II; Placental anticoagulant protein II; PP4-X; proliferation-inducing gene 28; proliferation-inducing protein 28; Protein II
Gene Aliases: ANX4; ANXA4; HEL-S-274; P32.5; PAP-II; PIG28; PP4-X; ZAP36
UniProt ID: (Human) P09525
Entrez Gene ID: (Human) 307
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.