R&D Systems
Manufacturer Code:MAB4855SP
Catalog # MAB4855SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 693510 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Annexin A3 Ala2-Asp323 Accession # P12429 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 35-alpha calcimedin; 35-alpha calcimedin annexin A312-cyclic-inositol-phosphate phosphodiesterase Annexin III annexin-3 ANXA3 Calcimedin 35-alpha Lipocortin III PAP-III Placental anticoagulant protein III placental anticoagulant protein III calcimedin 35-alpha; Annexin A3; Annexin III; Annexin III (lipocortin III); annexin III (lipocortin III, 1,2-cyclic-inositol-phosphate phosphodiesterase, placental anticoagulant protein III, calcimedin 35-alpha); Annexin-3; calcimedin 35-alpha; Inositol 1,2-cyclic phosphate 2-phosphohydrolase; Lipocortin III; PAP-III; Placental anticoagulant protein III
Gene Aliases: ANX3; ANXA3
UniProt ID: (Human) P12429
Entrez Gene ID: (Human) 306
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.