Novus Biologicals
Manufacturer Code:NBP234023
Catalog # NBP234023
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: DGVPKTEGDSSATALFPQTHKEQVQQDFSGKMQDLPEESSLEYQQEYFVTTPGTETSETQKAMIVPSSPSKTPEEVSTPAEEEKLYLQTPTSSERGGSPIIQEPEEPSEHREESSPRKTSLVIVESADNQPETCERLDEDAAFE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ANK-2; Ank2 Ankyrin 2 Ankyrin 2 brain Ankyrin B Ankyrin brain Ankyrin nonerythroid Brain Ankyrin; ankyrin B; ankyrin, brain; Ankyrin-2; ankyrin-2, nonerythrocytic; Ankyrin-B; Brain ankyrin; Non-erythroid ankyrin
Gene Aliases: ANK-2; ANK2; ANKB; brank-2; LQT4
UniProt ID: (Human) Q01484
Entrez Gene ID: (Human) 287
Molecular Function:
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.