Invitrogen
Manufacturer Code:MA536045
Catalog # PIMA536045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 001 |
Immunogen | Recombinant Mouse Angiotensinogen/AGT Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AGT AI265500 alpha-1 antiproteinase antitrypsin Ang Ang I Ang II Ang III Ang IV AngI AngII Angiotensin 1-10 Angiotensin 1-4 Angiotensin 1-5 Angiotensin 1-7 Angiotensin 1-8 Angiotensin 1-9 Angiotensin 2-8 Angiotensin 3-8 Angiotensin I Angiotensin II Angiotensin III Angiotensin IV angiotensin ll Angiotensin-1 Angiotensin-2 Angiotensin-3 Angiotensin-4 Angiotensinogen angiotensinogen (PAT) angiotensinogen (serpin peptidase inhibitor clade A member 8) ANHU ANRT Aogen Des-Asp[1]-angiotensin II PAT pre-angiotensinogen serine (or cysteine) proteinase inhibitor Serpin A8 serpin peptidase inhibitor clade A member 8 SERPINA8; angiotensin I; angiotensin ll; angiotensinogen; serpin A8
Gene Aliases: AI265500; AngI; AngII; Aogen; Serpina8
Entrez Gene ID: (Mouse) 11606
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.