Invitrogen
Manufacturer Code:PA578741
Catalog # PIPA578741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of mouse Angiotensinogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ang I; Ang II; Ang III; Ang IV; Angiotensin 1-10; Angiotensin 1-4; Angiotensin 1-5; Angiotensin 1-7; Angiotensin 1-8; Angiotensin 1-9; Angiotensin 2-8; Angiotensin 3-8; Angiotensin I; Angiotensin II; Angiotensin III; Angiotensin IV; angiotensin ll; Angiotensin-1; Angiotensin-2; Angiotensin-3; Angiotensin-4; Angiotensinogen; angiotensinogen (PAT); Angiotensinogen Serpin A8 Angiotensin-1 Angiotensin 1-10 Angiotensin I Ang I Angiotensin-2 Angiotensin 1-8 Angiotensin II Ang II Angiotensin-3 Angiotensin 2-8 Angiotensin III Ang III Des-Asp&lsqb1]-angiotensin II Angiotensin-4 Angiotensin 3-8 Angiotensin IV Ang IV Angiotensin 1-9 Angiotensin 1-7 Angiotensin 1-5; Des-Asp[1]-angiotensin II; Serpin A8
Gene Aliases: Agt; AI265500; Ang; AngI; AngII; ANRT; Aogen; PAT; Serpina8
UniProt ID: (Rat) P01015
Entrez Gene ID: (Rat) 24179, (Mouse) 11606
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.