Novus Biologicals
Manufacturer Code:NBP241247
Catalog # NBP241247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid peptide near the amino terminus of human Betatrophin. The immunogen is located within amino acids 40 - 90 of Betatrophin. Amino Acid Squence: VYRTTEGRLTKARNSL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: angiopoietin-like 8; Angiopoietin-like protein 8; Betatrophin; betatrophin variant 1; betatrophin variant 2; C19orf80 chromosome 19 open reading frame 80 hepatocellular carcinoma-associated gene TD26 hepatocellular carcinoma-associated protein TD26 PRO1185 PVPA599 TD26 UNQ599; hepatocellular carcinoma-associated gene TD26; hepatocellular carcinoma-associated protein TD26; Lipasin; Refeeding-induced fat and liver protein
Gene Aliases: ANGPTL8; C19orf80; PRO1185; PVPA599; RIFL; TD26; UNQ599/PRO1185
UniProt ID: (Human) Q6UXH0
Entrez Gene ID: (Human) 55908
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.