R&D Systems
Manufacturer Code:MAB7186SP
Catalog # MAB7186SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 748246 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant mouse Angiopoietin-2 Asp68-Phe496 Accession # O35608 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: AGPT2 ANG2 ANG-2 angiopoietin 2 angiopoietin-2 angiopoietin-2a angiopoietin-2B angiopoitin 2 ANGPT2 Tie2-ligand; ANG-2; Angiopoietin-2; angiopoietin-2a; angiopoietin-2B; Tie2-ligand
Gene Aliases: AGPT2; ANG2; ANGPT2
UniProt ID: (Human) O15123
Entrez Gene ID: (Human) 285
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.