Novus Biologicals
Manufacturer Code:NBP289600100UL
Catalog # NB020683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 117 |
Immunogen | Recombinant Human Angiopoietin-2 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AGPT2 ANG2 ANG-2 angiopoietin 2 angiopoietin-2 angiopoietin-2a angiopoietin-2B angiopoitin 2 Tie2-ligand; ANG-2; Angiopoietin-2; angiopoietin-2a; angiopoietin-2B; Tie2-ligand
Gene Aliases: AGPT2; ANG2; ANGPT2
UniProt ID: (Human) O15123
Entrez Gene ID: (Human) 285
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.