Invitrogen
Manufacturer Code:PA546968
Catalog # PIPA546968
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit human Angiopoietin-1 Ser20-Phe498 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: AGP1 AGPT ANG1 ANG-1 angiopoietin-1; ANG-1; Angiopoietin-1
Gene Aliases: 1110046O21Rik; AGP1; AGPT; Ang-1; ANG1; ANGPT1; KIAA0003
UniProt ID: (Human) Q15389, (Mouse) O08538
Entrez Gene ID: (Human) 284, (Mouse) 11600
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.