Invitrogen
Manufacturer Code:PA132150
Catalog # PIPA132150
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to the N-terminus residues CN(21) Q R R N P E N G G R R Y N R I Q H G Q(40) of mouse Ang-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110046O21Rik Ang-1 Ang1; ANG-1; Angiopoietin-1
Gene Aliases: 1110046O21Rik; Agpt; Ang-1; Ang1; Angpt1
UniProt ID: (Mouse) O08538
Entrez Gene ID: (Mouse) 11600
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.