Proteintech
Manufacturer Code:183021AP150UL
Catalog # 50-173-2101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALS9 ANG Angiogenin HEL168 Ribonuclease 5 RNase 5 RNASE4 RNASE5; Angiogenin; angiogenin, ribonuclease, RNase A family, 5; epididymis luminal protein 168; Ribonuclease 5; ribonuclease A A1; ribonuclease A family member 5; RNase 5
Gene Aliases: ALS9; ANG; HEL168; RAA1; RNASE4; RNASE5
UniProt ID: (Human) P03950
Entrez Gene ID: (Human) 283
Molecular Function: RNA binding protein endoribonuclease enzyme modulator hydrolase nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.