Proteintech
Manufacturer Code:158451AP150UL
Catalog # 50-173-0524
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amylase alpha Fusion Protein Ag8605 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; AMY2A Amylase alpha Pancreatic alpha amylase; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; glycogenase; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Aliases: Amy-X; AMY2; Amy2-1; Amy2-2; AMY2A; mAmy2-1; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022462; PA
UniProt ID: (Human) Q9UBH3
Entrez Gene ID: (Human) 279, (Mouse) 100043207, (Mouse) 545562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.