Proteintech
Manufacturer Code:125401AP150UL
Catalog # 50-172-8050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Amylase alpha Fusion Protein Ag3260 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 2B; Alpha amylase 2B AMY2 AMY2B Amylase alpha amylase alpha 2B (pancreatic) Carcinoid alpha amylase; Alpha-amylase 2B; amylase 2-2, pancreatic; amylase 2b; amylase 2b, pancreatic; Carcinoid alpha-amylase; glycogenase
Gene Aliases: Amy-X; AMY2; Amy2-2; AMY2B; AMY3; HXA; mAmy2-1; OTTMUSG00000022462
UniProt ID: (Human) P19961
Entrez Gene ID: (Human) 280, (Mouse) 545562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.