Invitrogen
Manufacturer Code:25537041
Catalog # 25-537-041
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AREG559 |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Amphiregulin; amphiregulin B; AR; Colorectum cell-derived growth factor; Colorectum cell-derived growth factor CRDGF AREG SDGF Mcub AREGB MGC13647; CRDGF; schwannoma-derived growth factor
Gene Aliases: AR; AREG; AREGB; CRDGF; SDGF
UniProt ID: (Human) P15514
Entrez Gene ID: (Human) 374
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.