Invitrogen
Manufacturer Code:PA547903
Catalog # PIPA547903
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit mouse Amnionless Ala20-Gly362 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AMN amnionless homolog (mouse) PRO1028 protein amnionless UNQ513/PRO1028 visceral endoderm-specific type 1 transmembrane protein; Protein amnionless; Soluble protein amnionless; type I transmembrane protein
Gene Aliases: 5033428N14Rik; Amn; AV002116
UniProt ID: (Mouse) Q99JB7
Entrez Gene ID: (Mouse) 93835
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.