Invitrogen
Manufacturer Code:PA551960
Catalog # PIPA551960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Amisyn |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Amisyn; STXBP6 syntaxin binding protein 6 (amisyn) amisyn HSPC156 amisyn syntaxin binding protein 6 FLJ39638 OTTHUMP00000178754; syntaxin binding protein 6 (amisyn); Syntaxin-binding protein 6
Gene Aliases: amisyn; HSPC156; STXBP6
UniProt ID: (Human) Q8NFX7
Entrez Gene ID: (Human) 29091
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.