Novus Biologicals
Manufacturer Code:NBP270171F
Catalog # NBP270171F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6F4 |
Immunogen | Recombinant protein fragment corresponding to amino acids 146-403 of human Aminomethyltransferase (NP_000472) produced in E.coli. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: aminomethyltransferase EC 2.1.2.10 GCE GCSTaminomethyltransferase (glycine cleavage system protein T) glycine cleavage system protein T NKHmitochondrial; Aminomethyltransferase, mitochondrial; GCVT; Glycine cleavage system T protein
Gene Aliases: AMT; GCE; GCST; GCVT; NKH
UniProt ID: (Human) P48728
Entrez Gene ID: (Human) 275
Molecular Function: dehydrogenase oxidase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.