Novus Biologicals
Manufacturer Code:NBP28955950UL
Catalog # NB020602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 128 |
Immunogen | Recombinant Human Aminoacylase/ACY1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ACY-1; ACY-1 ACY1D acylase aminoacylase 1 aminoacylase-1 EC 3.5.1 EC 3.5.1.14 N-acyl-L-amino-acid amidohydrolase; acylase; Aminoacylase-1; epididymis secretory protein Li 5; N-acyl-L-amino-acid amidohydrolase
Gene Aliases: ACY-1; ACY1; ACY1D; HEL-S-5
UniProt ID: (Human) Q03154
Entrez Gene ID: (Human) 95
Molecular Function: deacetylase hydrolase metalloprotease protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.