R&D Systems
Manufacturer Code:MAB9830100
Catalog # MAB9830100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2285D |
Immunogen | Synthetic peptide containing human ACTN2 Accession # P35609 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit Monoclonal Antibody
Protein Aliases: Actinin alpha 2 actinin alpha 2 Actinin-a2 ACTN2 alpha-Actinin 2 Alpha-Actinin Skeletal Muscle Alpha-actinin skeletal muscle isoform 2 alpha-actinin-2 CMD1AA F-actin cross-linking protein; actinin, alpha 2; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 2; Alpha-actinin-2; F-actin cross-linking protein
Gene Aliases: ACTN2; CMD1AA; CMH23
UniProt ID: (Human) P35609
Entrez Gene ID: (Human) 88
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.