Novus Biologicals
Manufacturer Code:NBP29217020UL
Catalog # NB026232
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human TTPA (NP_000361.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-tocopherol transfer protein; alpha-tocopherol transfer protein alphaTTP Alpha-TTP ataxia (Friedreich-like) with vitamin E deficiency ATTP AVED tocopherol (alpha) transfer protein tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin Edeficiency) TPP1 TTP1; Alpha-TTP; tocopherol (alpha) transfer protein (ataxia (Friedreich-like) with vitamin E deficiency)
Gene Aliases: alphaTTP; ATTP; AVED; TPP1; TTP1; TTPA
UniProt ID: (Human) P49638
Entrez Gene ID: (Human) 7274
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.