Invitrogen
Manufacturer Code:MA511544
Catalog # PIMA511544
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1A4 (asm-1) |
Immunogen | N-terminal decapeptide of alpha-smooth muscle isoform of actin acetylated at the N-terminus |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: actin aortic smooth muscle alpha 2 actin alpha-actin-2 alpha-cardiac actin cell growth-inhibiting gene 46 protein growth-inhibiting gene 46 alpha-sma; actin, alpha 2, smooth muscle, aorta; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; alpha actin 2; Alpha-actin; Alpha-actin-2; alpha-cardiac actin; alpha-smooth muscle actin; Cell growth-inhibiting gene 46 protein; smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AAT6; ACTA2; ACTSA; ACTVS; alphaSMA; asma; GIG46; MYMY5; SMalphaA
UniProt ID: (Rabbit) P62740, (Chicken) P08023, (Bovine) P62739, (Human) P62736, (Rat) P62738, (Mouse) P62737
Entrez Gene ID: (Rabbit) 100009271, (Chicken) 423787, (Bovine) 515610, (Human) 59, (Rat) 81633, (Mouse) 11475
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.