Invitrogen
Manufacturer Code:MA536194
Catalog # PIMA536194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JE54-66 |
Immunogen | Synthetic peptide within C-terminal Human Alpha SNAP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 1500039N14Rik alpha snap alpha soluble NSF attachment protein alpha-SNAP Alpha-soluble NSF attachment protein a-SNAP AW209189 hydrocephaly with hop gait hyh NAPA N-ethylmaleimide sensitive fusion protein attachment protein alpha N-ethylmaleimide-sensitive factor attachment protein alpha N-ethylmaleimide-sensitive factor attachment protein alpha NSF attachment protein alpha RA81 Snap SNAPA SNAP-alpha SNARE; alpha soluble NSF attachment protein; alpha-SNAP; Alpha-soluble NSF attachment protein; hydrocephaly with hop gait; N-ethylmaleimide-sensitive factor attachment protein alpha; N-ethylmaleimide-sensitive factor attachment protein, alpha; SNAP-alpha
Gene Aliases: 1500039N14Rik; a-SNAP; AW209189; hyh; NAPA; RA81; SNAPA; SNARE
UniProt ID: (Human) P54920, (Mouse) Q9DB05
Entrez Gene ID: (Human) 8775, (Mouse) 108124
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.