Proteintech
Manufacturer Code:551351AP150UL
Catalog # 50-173-6008
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a SMA AAT6 ACTA2 ACTA2 Smooth muscle Actin ACTA2 Smooth muscle Actin Specific Actin aortic smooth muscle Alpha actin 2 Alpha SMA Alpha SMA Specific a-SMA smooth muscle actin alpha SMA; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; Alpha-actin-2; alpha-cardiac actin; Cell growth-inhibiting gene 46 protein; smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AAT6; ACTA2; ACTSA; ACTVS; alphaSMA; GIG46; MYMY5; SMalphaA
UniProt ID: (Human) P62736, (Rat) P62738, (Mouse) P62737
Entrez Gene ID: (Human) 59, (Rat) 81633, (Mouse) 11475
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.