Invitrogen
Manufacturer Code:PA578771
Catalog # PIPA578771
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human Alpha Amylase 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 1; alpha amylase 1; Alpha-amylase 1; Alpha-amylase 1A; Alpha-amylase 1B; Alpha-amylase 1C; amylase 1, salivary; amylase, salivary, alpha-1A; glycogenase; Salivary alpha-amylase; salivary amylase alpha 1A; Salivary and hepatic alpha-amylase
Gene Aliases: Amy-1; Amy-1-a; AMY1; AMY1A; AMY1B; AMY1C; C030014B17Rik
UniProt ID: (Human) P0DUB6, (Mouse) P00687
Entrez Gene ID: (Human) 276, (Mouse) 11722, (Rat) 24203
Molecular Function:
amylase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.