Novus Biologicals
Manufacturer Code:NBP215358
Catalog # NBP215358
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human alpha Actinin 4. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: actinin alpha4 isoform actinin alpha 4 ACTININ-4 alpha-actinin-4 DKFZp686K23158 F-actin cross-linking protein focal segmental glomerulosclerosis 1 FSGS FSGS1 Non-muscle alpha-actinin 4; Alpha-actinin-4; focal segmental glomerulosclerosis 1; Non-muscle alpha-actinin 4
Gene Aliases: ACTININ-4; ACTN4; FSGS; FSGS1
UniProt ID: (Human) O43707
Entrez Gene ID: (Human) 81
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.