Invitrogen
Manufacturer Code:MA526164
Catalog # PIMA526164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B12 |
Immunogen | Full length human recombit protein of CRYAA produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acry 1 Alpha crystallin A chain Alpha-crystallin A chain CRYA 1 CRYA1 CRYAA CRYAA&lowbarHUMAN Crystallin Alpha 1 Crystallin alpha A Heat shock protein beta 4 Heat shock protein beta-4 HSPB 4 HspB4 short form Zonular Central Nuclear Cataract; alpha crystallin A chain; Alpha-crystallin A chain; Alpha-crystallin A(1-162); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); crystallin, alpha A; crystallin, alpha-1; Heat shock protein beta-4; HspB4; human alphaA-crystallin (CRYA1)
Gene Aliases: CRYA1; CRYAA; CRYAA2; CTRCT9; HSPB4
UniProt ID: (Human) A0A140G945
Entrez Gene ID: (Human) 1409
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.