Invitrogen
Manufacturer Code:PA576806
Catalog # PIPA576806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CRYAA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acry 1 Alpha crystallin A chain Alpha-crystallin A chain CRYA 1 CRYA1 CRYAA CRYAA&lowbarHUMAN Crystallin Alpha 1 Crystallin alpha A Heat shock protein beta 4 Heat shock protein beta-4 HSPB 4 HspB4 short form Zonular Central Nuclear Cataract; alpha crystallin A chain; Alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; Heat shock protein beta-4; HspB4; human alphaA-crystallin (CRYA1); lens opacity 18
Gene Aliases: Acry-1; Crya-1; CRYA1; CRYAA; CRYAA2; CTRCT9; DAcry-1; HSPB4; lop18
UniProt ID: (Human) A0A140G945, (Rat) P24623, (Mouse) P24622
Entrez Gene ID: (Human) 1409, (Rat) 24273, (Mouse) 12954
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.