Invitrogen
Manufacturer Code:PA5114152
Catalog # PIPA5114152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal region of Human ADRA2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADRA2 Adra-2 ADRA2A Adra-2a ADRA2R ADRAR adrenergic receptor alpha 2a adrenergic alpha-2A- receptor adrenoceptor alpha 2A alpha(2A)AR Alpha-2 adrenergic receptor subtype C10 alpha2A Alpha-2A adrenergic receptor Alpha-2A adrenoceptor Alpha-2A adrenoreceptor alpha2A-adrenergic receptor ALPHA2AAR alpha-2AAR alpha2A-AR alpha-2AAR subtype C10 alpha-2-adrenergic receptor platelet type alpha2-C10 Alpha-2D adrenergic receptor AW122659 CA2-47 OTTHUMP00000058849 RATRG20 RG20 subtype alpha2-C10 ZNF32; adrenergic, alpha-2A-, receptor; Alpha-2 adrenergic receptor subtype C10; alpha-2-adrenergic receptor, platelet type; Alpha-2A adrenergic receptor; Alpha-2A adrenoceptor; Alpha-2A adrenoreceptor; alpha-2AAR subtype C10
Gene Aliases: ADRA2; ADRA2A; ADRA2R; ADRAR; ALPHA2AAR; ZNF32
UniProt ID: (Human) P08913
Entrez Gene ID: (Human) 150
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.