Invitrogen
Manufacturer Code:PA547920
Catalog # PIPA547920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human Allergin-1 (isoform Allergin-1S2) Met29-Ser131 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Allergin-1; Allergy inhibitory receptor 1; C17orf60 MCA32 MCA32-1S2 MILR1 Allergin-1 Allergy Inhibitory Receptor 1 C17orf60 Chromosome 17 Open Reading Frame 60 Mast Cell Antigen 32 Mast Cell Immunoglobulin-Like Receptor 1 MCA32 MCA-32 Probable Mast Cell Antigen 32 Homolog; Mast cell antigen 32; Mast cell immunoglobulin-like receptor 1; MCA-32; probable mast cell antigen 32 homolog
Gene Aliases: Allergin-1; C17orf60; MCA-32; MCA32; MILR1
UniProt ID: (Human) Q7Z6M3
Entrez Gene ID: (Human) 284021
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.