Invitrogen
Manufacturer Code:PA5113085
Catalog # PIPA5113085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Allergin-1 protein (249-343AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Allergin-1; Allergin-1 Allergy inhibitory receptor 1 C17orf60 Mast cell antigen 32 mast cell immunoglobulin like receptor 1 Mast cell immunoglobulin-like receptor 1 MCA32 MCA-32 MCA32-1S2 MILR1 probable mast cell antigen 32 homolog; Allergy inhibitory receptor 1; mast cell Ag-32; Mast cell antigen 32; Mast cell immunoglobulin-like receptor 1; MCA-32; probable mast cell antigen 32 homolog; surface protein MCA-32
Gene Aliases: Allergin-1; C17orf60; Gm885; MCA-32; MCA32; MILR1
UniProt ID: (Human) Q7Z6M3, (Mouse) Q3TB92
Entrez Gene ID: (Human) 284021, (Mouse) 380732
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.