Novus Biologicals
Manufacturer Code:NBP267295
Catalog # NBP267295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SA40-00 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase liver/bone/kidney isozyme alkaline phosphatase liver/bone/kidney alkaline phosphatase tissue-nonspecific isozyme alkaline phosphomonoesterase APTNAP AP-TNAP EC 3.1.3.1 FLJ40094 FLJ93059 glycerophosphatase HOPS liver/bone/kidney-type alkaline phosphatase MGC161443 tissue-nonspecific ALP TNAP TNSALPMGC167935; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific ALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.