Novus Biologicals
Manufacturer Code:NBP222193
Catalog # NBP222193
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2F4 |
Immunogen | Purified recombinant fragment of human Alkaline Phosphatase Tissue Non-Specific expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase liver/bone/kidney isozyme alkaline phosphatase liver/bone/kidney alkaline phosphatase tissue-nonspecific isozyme alkaline phosphomonoesterase APTNAP AP-TNAP EC 3.1.3.1 FLJ40094 FLJ93059 glycerophosphatase HOPS liver/bone/kidney-type alkaline phosphatase MGC161443 tissue-nonspecific ALP TNAP TNSALPMGC167935; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific ALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.