R&D Systems
Manufacturer Code:MAB29091
Catalog # MAB29091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 388828 |
Immunogen | <em>E. coli</em>-derived recombinant human Alkaline Phosphatase/ALPL Leu18-Ser502 Accession # P05186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Akp2 Alkaline phosphatase liver/bone/kidney isozyme alkaline phosphatase liver/bone/kidney alkaline phosphatase tissue-nonspecific isozyme alkaline phosphomonoesterase APTNAP AP-TNAP EC 3.1.3.1 FLJ40094 FLJ93059 glycerophosphatase HOPS liver/bone/kidney-type alkaline phosphatase MGC161443 tissue-nonspecific ALP TNAP TNSALP TNSALPMGC167935; Alkaline phosphatase liver/bone/kidney isozyme; Alkaline phosphatase, tissue-nonspecific isozyme; AP-TNAP; liver/bone/kidney-type alkaline phosphatase; tissue-nonspecific ALP
Gene Aliases: ALPL; AP-TNAP; APTNAP; HOPS; TNAP; TNSALP
UniProt ID: (Human) P05186
Entrez Gene ID: (Human) 249
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.