Novus Biologicals
Manufacturer Code:NB10065678JF646
Catalog # NB065678646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3A9 |
Immunogen | Alix fusion protein |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: AIP1DRIP4 ALG-2 interacting protein 1 ALG-2 interacting protein X ALG-2-interacting protein 1 alinx ALIX apoptosis-linked gene 2-interacting protein X dopamine receptor interacting protein 4 HP95 KIAA1375 MGC17003 PDCD6-interacting protein programmed cell death 6 interacting protein programmed cell death 6-interacting protein; ALG-2 interacting protein 1; ALG-2-interacting protein 1; ALG-2-interacting protein X; apoptosis-linked gene 2-interacting protein X; dopamine receptor interacting protein 4; Hp95; PDCD6-interacting protein; Programmed cell death 6-interacting protein
Gene Aliases: AIP1; ALIX; DRIP4; HP95; KIAA1375; PDCD6IP
UniProt ID: (Human) Q8WUM4
Entrez Gene ID: (Human) 10015
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.