Invitrogen
Manufacturer Code:PA588178
Catalog # PIPA588178
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-170 of human ALDOC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; Brain-type aldolase; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; Fructose-bisphosphate aldolase C; Scrapie-responsive protein 2; Zebrin II
Gene Aliases: AI847350; ALDC; ALDCAA; Aldo3; ALDOC; AU040929; F16dip7; RATALDCAA; Scrg2
UniProt ID: (Human) P09972, (Mouse) P05063, (Rat) P09117
Entrez Gene ID: (Human) 230, (Mouse) 11676, (Rat) 24191
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.