Novus Biologicals
Manufacturer Code:NBP242620AF350
Catalog # NP242620A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F9 |
Immunogen | Recombinant human ALDOA (1-364aa) purified from E. coli |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ALDA aldolase A fructose-bisphosphate EC 4.1.2.13 fructose-16-bisphosphate triosephosphate-lyase fructose-bisphosphate aldolase A GSD12 Lung cancer antigen NY-LU-1 MGC10942 MGC17716 MGC17767 Muscle-type aldolase; aldolase A, fructose-bisphosphate; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; Fructose-bisphosphate aldolase A; Lung cancer antigen NY-LU-1; Muscle-type aldolase
Gene Aliases: ALDA; ALDOA; GSD12; HEL-S-87p
UniProt ID: (Human) P04075
Entrez Gene ID: (Human) 226
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.