Invitrogen
Manufacturer Code:PA577857
Catalog # PIPA577857
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Aldolase A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALDOA aldolase A fructose-bisphosphate ALDA GSD12; Aldolase 1; aldolase 1, A isoform; aldolase A, fructose-bisphosphate; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; Fructose-bisphosphate aldolase A; Lung cancer antigen NY-LU-1; Muscle-type aldolase
Gene Aliases: ALDA; Aldo-1; Aldo1; ALDOA; GSD12; HEL-S-87p; RNALDOG5
UniProt ID: (Human) P04075, (Mouse) P05064, (Rat) P05065
Entrez Gene ID: (Human) 226, (Mouse) 11674, (Rat) 24189
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.