Novus Biologicals
Manufacturer Code:NBP297028R
Catalog # NB080730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Aldo-keto Reductase 1C4/AKR1C4 (AAH20744.1 Met1-Tyr323). |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-alpha-HSD AKR1C4 aldo-keto reductase family 1 member C4 C11 CDR CHDR DD4 DD-4 HAKRA; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; 3alpha-hydroxysteroid 3-dehydrogenase; Aldo-keto reductase family 1 member C4; CDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; type I 3-alpha-hydroxysteroid dehydrogenase
Gene Aliases: 3-alpha-HSD; AKR1C4; C11; CDR; CHDR; DD-4; DD4; HAKRA
UniProt ID: (Human) P17516
Entrez Gene ID: (Human) 1109
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.