R&D Systems
Manufacturer Code:AF6957
Catalog # AF6957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Aldo-keto Reductase 1C4/AKR1C4 Met1-Tyr323 Accession # NP_001809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-alpha-HSD AKR1C4 aldo-keto reductase family 1 member C4 AldoketoReductase 1C4 C11 CDR CHDR DD4 DD-4 HAKRA; 3-alpha-HSD1; 3-alpha-hydroxysteroid dehydrogenase type I; 3alpha-hydroxysteroid 3-dehydrogenase; Aldo-keto reductase family 1 member C4; CDR; Chlordecone reductase; chlordecone reductase; 3-alpha hydroxysteroid dehydrogenase, type I; dihydrodiol dehydrogenase 4; DD-4; Dihydrodiol dehydrogenase 4; dihydrodiol dehydrogenase isozyme DD4; HAKRA; type I 3-alpha-hydroxysteroid dehydrogenase
Gene Aliases: 3-alpha-HSD; AKR1C4; C11; CDR; CHDR; DD-4; DD4; HAKRA
UniProt ID: (Human) P17516
Entrez Gene ID: (Human) 1109
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.