R&D Systems
Manufacturer Code:AF7678SP
Catalog # AF7678SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Aldo-keto Reductase 1C3/AKR1C3 Met1-Tyr323 Accession # P42330 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 5; 17-beta-hydroxysteroid dehydrogenase type 5; 17-beta-hydroxysteroid dehydrogenase type 5 3-alpha-HSD type 2 3-alpha-hydroxysteroid dehydrogenase type 2 AKR1C3 aldo-keto reductase family 1 member C3 aldo-keto reductase family 1 member C3 (3-alpha hydroxysteroid dehydrogenase type II) AldoketoReductase 1C3 Chlordecone reductase homolog HAKRb DD3 DD-3 DD317-beta-HSD 5 DDH1 DDX Dihydrodiol dehydrogenase 3 Dihydrodiol dehydrogenase type I dihydrodiol dehydrogenase X EC 1.1.1 EC 1.1.1.1123-alpha-HSD type II brain EC 1.1.1.188 EC 1.1.1.213 EC 1.1.1.63 EC 1.1.1.64 EC 1.3.1.20 HA1753 HAKRB HAKRBtype IIb 3-alpha hydroxysteroid dehydrogenase HAKRe HSD17B5 hydroxysteroid (17-beta) dehydrogenase 5 Indanol dehydrogenase KIAA0119hluPGFS PGFS Prostaglandin F synthase Testosterone 17-beta-dehydrogenase 5 Trans-12-dihydrobenzene-12-diol dehydrogenase; 3-alpha hydroxysteroid dehydrogenase, type II; 3-alpha-HSD type 2; 3-alpha-HSD type II, brain; 3-alpha-hydroxysteroid dehydrogenase type 2; Aldo-keto reductase family 1 member C3; Chlordecone reductase homolog HAKRb; DD-3; Dihydrodiol dehydrogenase 3; Dihydrodiol dehydrogenase type I; dihydrodiol dehydrogenase X; HA1753; indanol dehydrogenase; PGFS; Prostaglandin F synthase; Testosterone 17-beta-dehydrogenase 5; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type IIb 3-alpha hydroxysteroid dehydrogenase
Gene Aliases: AKR1C3; DD3; DDH1; DDX; HA1753; HAKRB; HAKRe; hluPGFS; HSD17B5; KIAA0119; PGFS
UniProt ID: (Human) P42330
Entrez Gene ID: (Human) 8644
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.