Invitrogen
Manufacturer Code:PA520603
Catalog # PIPA520603
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide near the carboxy terminus of the human Aldh3A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde dehydrogenase 10; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A2; Aldehyde dehydrogenase family 3 member A2; ALDH10 FALDH SLS aldehyde dehydrogenase 10 aldehyde dehydrogenase family 3 member A2 fatty aldehyde dehydrogenase microsomal aldehyde dehydrogenase; Fatty aldehyde dehydrogenase; Microsomal aldehyde dehydrogenase
Gene Aliases: Ahd-3; Ahd-3r; Ahd3; Ahd3-r; AI194803; ALDH10; Aldh3; ALDH3A2; Aldh4; Aldh4-r; FALDH; SLS
UniProt ID: (Human) P51648, (Mouse) P47740
Entrez Gene ID: (Human) 224, (Mouse) 11671
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.