Invitrogen
Manufacturer Code:PA520602
Catalog # PIPA520602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 13 amino acid peptide near the carboxy terminus of the human Aldh3A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alcohol dehydrogenase family 3, subfamily A1; Aldehyde dehydrogenase (Ahd-c); Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A1; aldehyde dehydrogenase 3, stomach cytosolic (class 3); aldehyde dehydrogenase 3A1; Aldehyde dehydrogenase 4; aldehyde dehydrogenase class 3; Aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase family 3 subfamily A1; aldehyde dehydrogenase family 3, member A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; Aldehyde dehydrogenase, dimeric NADP-preferring; ALDH3 ALDHIII aldehyde dehydrogenase isozyme 3 aldehyde dehydrogenase type III aldehyde dehydrogenase dimeric NADP-preferring stomach aldehyde dehydrogenase; ALDHIII; Dioxin-inducible aldehyde dehydrogenase 3; HTC-ALDH; stomach aldehyde dehydrogenase; Tumor-associated aldehyde dehydrogenase; tumor-associated aldehyde dehydrogenase tumor ALDH
Gene Aliases: Ahd-4; Ahd-c; Ahd4; AHDC; Aldd; Aldh; ALDH3; ALDH3A1; Aldh4; ALDHIII
UniProt ID: (Human) P30838, (Mouse) P47739, (Rat) P11883
Entrez Gene ID: (Human) 218, (Mouse) 11670, (Rat) 25375
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.