Abnova
Manufacturer Code:PAB5866
Catalog # 89-131-960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Aldh1l1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Aldh1l1.
For Research Use Only
Protein Aliases: 10-formyltetrahydrofolate dehydrogenase; 10-FTHFDH; 1810048F20Rik; Aldehyde dehydrogenase family 1 member L1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; formyltetrahydrofolate dehydrogenase; Fthfd
Gene Aliases: 1810048F20Rik; Aldh1l1; FDH; Fthfd; Neut2
UniProt ID: (Mouse) Q8R0Y6
Entrez Gene ID: (Mouse) 107747
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.