Novus Biologicals
Manufacturer Code:NBP292428100UL
Catalog # NB026726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 340-590 of human AOX1 (NP_001150.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde oxidase; Aldehyde oxidase 1; aldehyde oxidase aldehyde oxidase 1 AOAOH1 EC 1.2.3.1; Azaheterocycle hydroxylase
Gene Aliases: AO; AOH1; AOX1
UniProt ID: (Human) Q06278
Entrez Gene ID: (Human) 316
Molecular Function:
dehydrogenase
oxidase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.