Novus Biologicals
Manufacturer Code:NBP289893FR
Catalog # NB109957
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 23 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 (NP_000680.2 Ser 2-Ser 501). |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 1; acetaldehyde dehydrogenase 1 ALDC aldehyde dehydrogenase 1 family member A1 aldehyde dehydrogenase 1 soluble Aldehyde dehydrogenase family 1 member A1 Aldehyde dehydrogenase cytosolic aldehyde dehydrogenase liver cytosolic ALDH class 1 ALDH1 ALDH11 ALDH-E1 ALHDII EC 1.2.1 EC 1.2.1.36 MGC2318 PUMB1 RALDH 1 RalDH1 retinal dehydrogenase 1 retinaldehyde dehydrogenase 1; aldehyde dehydrogenase 1 family, member A1; aldehyde dehydrogenase 1, soluble; Aldehyde dehydrogenase family 1 member A1; Aldehyde dehydrogenase, cytosolic; aldehyde dehydrogenase, liver cytosolic; ALDH class 1; ALDH-E1; ALHDII; epididymis luminal protein 12; epididymis luminal protein 9; epididymis secretory sperm binding protein Li 53e; RALDH 1; Retinal dehydrogenase 1; retinaldehyde dehydrogenase 1
Gene Aliases: ALDC; ALDH-E1; ALDH1; ALDH11; ALDH1A1; HEL-9; HEL-S-53e; HEL12; PUMB1; RALDH1
UniProt ID: (Human) P00352
Entrez Gene ID: (Human) 216
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.