Invitrogen
Manufacturer Code:PA518332
Catalog # PIPA518332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (STAGKVMKCKA) corresponding to the N-terminus amino acids of ADH1A B C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; ADH1A ADH1B ADH1C pan ADH ADH1B ADH2 alcohol dehydrogenase IB ADH beta aldehyde reductase alcohol dehydrogenase 2 ADH gamma alcohol dehydrogenase 3 class I gamma polypeptide dehydrogenase 2 class I alcohol dehydrogenase 1B class I ADH1C alcohol dehydrogenase 1C class I ADH3 aldehyde reductase class I alcohol dehydrogenase ADH1A alcohol dehydrogenase 1A; alcohol dehydrogenase 1 (class I), alpha polypeptide; Alcohol dehydrogenase 1A; Alcohol dehydrogenase 1B; Alcohol dehydrogenase 1C; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; Alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117
Gene Aliases: ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; HEL-S-117
UniProt ID: (Human) P07327, (Human) P00325, (Human) P00326
Entrez Gene ID: (Human) 124, (Human) 125, (Human) 126
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.