Novus Biologicals
Manufacturer Code:NBP298272AF700
Catalog # NB098057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Alanyl tRNA synthetase (Q8BGQ7 Met1-Asn968). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AARS; alanine tRNA ligase 1 cytoplasmic Alanine--tRNA ligase alanyl-tRNA synthetase alanyl-tRNA synthetase cytoplasmic AlaRS CMT2N EC 6.1.1 EC 6.1.1.7 Renal carcinoma antigen NY-REN-42; alanine tRNA ligase 1, cytoplasmic; Alanine--tRNA ligase, cytoplasmic; Alanyl-tRNA synthetase; alanyl-tRNA synthetase, cytoplasmic; AlaRS; Renal carcinoma antigen NY-REN-42
Gene Aliases: AARS; AARS1; CMT2N; EIEE29
UniProt ID: (Human) P49588
Entrez Gene ID: (Human) 16
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.