Abnova
Manufacturer Code:PAB5897
Catalog # 89-131-988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Aicda. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Aicda.
For Research Use Only
Protein Aliases: activation induced deaminase; Activation-induced cytidine deaminase; AID; Arp2; Cytidine aminohydrolase; Single-stranded DNA cytosine deaminase
Gene Aliases: Aicda; Aid; Arp2
UniProt ID: (Mouse) Q9WVE0
Entrez Gene ID: (Mouse) 11628
Molecular Function:
deaminase
hydrolase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.