R&D Systems
Manufacturer Code:MAB550SP
Catalog # MAB550SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 76805 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant rat Agrin Ala1153-Pro1959 (Pro1788-Ser1798 del) Accession # P25304 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Agrin; agrin agrin proteoglycan AGRN; Agrin C-terminal 110 kDa subunit; Agrin C-terminal 22 kDa fragment; Agrin C-terminal 90 kDa fragment; Agrin N-terminal 110 kDa subunit; agrin proteoglycan; C22; C90
Gene Aliases: AGRIN; AGRN; CMS8; CMSPPD
UniProt ID: (Human) O00468
Entrez Gene ID: (Human) 375790
Molecular Function:
extracellular matrix linker protein
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.